Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204840.1 | NP_001191769.1 | 2 - 3 | 2181 | 76 | |
| XM_008980717.1 | XP_008978965.1 | 3 - 4 | 2258 | 76 | ||
| XM_008980718.2 | XP_008978966.1 | 3 - 4 | 2934 | 76 | ||
More Information
Set Membership: |
|

STON1
Genomic Map