Assay Details
| Gene Symbol: MTRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270912.1 | NP_001257841.1 | 2 - 3 | 519 | 96 | |
| XM_008998095.2 | XP_008996343.1 | 2 - 3 | 471 | 96 | ||
| XM_008998096.2 | XP_008996344.1 | 3 - 4 | 1150 | 96 | ||
| XM_008998097.2 | XP_008996345.1 | 2 - 3 | 669 | 96 | ||
More Information
Set Membership: |
|

MTRF1
Genomic Map