Assay Details
| Gene Symbol: MTRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270912.1 | NP_001257841.1 | 8 - 9 | 1229 | 68 | |
| XM_008998095.2 | XP_008996343.1 | 8 - 9 | 1181 | 68 | ||
| XM_008998096.2 | XP_008996344.1 | 9 - 10 | 1860 | 68 | ||
| XM_008998097.2 | XP_008996345.1 | 8 - 9 | 1379 | 68 | ||
More Information
Set Membership: |
|

MTRF1
Genomic Map