Assay Details
| Gene Symbol: LDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755086.4 | XP_002755132.4 | 1 - 2 | 359 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LDHA| Gene Symbol: LDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755086.4 | XP_002755132.4 | 1 - 2 | 359 | 69 | |
Set Membership: |
|