Assay Details
| Gene Symbol: LIPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761316.3 | XP_002761362.3 | 3 - 4 | 574 | 99 | |
| XM_008986482.2 | XP_008984730.1 | 3 - 4 | 574 | 99 | ||
| XM_008986483.2 | XP_008984731.1 | 3 - 4 | 574 | 99 | ||
| XM_008986484.1 | XP_008984732.1 | 3 - 4 | 574 | 99 | ||
| XM_008986485.2 | XP_008984733.1 | 3 - 4 | 574 | 99 | ||
More Information
Set Membership: |
|

LIPI
Genomic Map