Assay Details
| Gene Symbol: RIPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733973.3 | XP_003734021.2 | 9 - 10 | 1999 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIPK3| Gene Symbol: RIPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733973.3 | XP_003734021.2 | 9 - 10 | 1999 | 66 | |
Set Membership: |
|