Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981504.1 | XP_008979752.1 | 2 - 3 | 850 | 94 | |
| XM_008981505.1 | XP_008979753.1 | 2 - 3 | 850 | 94 | ||
| XM_008981506.2 | XP_008979754.1 | 2 - 3 | 850 | 94 | ||
More Information
Set Membership: |
|

ZKSCAN7
Genomic Map