Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981504.1 | XP_008979752.1 | 5 - 6 | 1241 | 76 | |
| XM_008981506.2 | XP_008979754.1 | 5 - 6 | 1241 | 76 | ||
More Information
Set Membership: |
|

ZKSCAN7
Genomic Map