Assay Details
| Gene Symbol: HMHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965020.1 | XP_017820509.1 | 2 - 3 | 106 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMHB1| Gene Symbol: HMHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965020.1 | XP_017820509.1 | 2 - 3 | 106 | 97 | |
Set Membership: |
|