Assay Details
| Gene Symbol: LOC100414470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744278.3 | XP_002744324.1 | 1 | 4122 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100414470| Gene Symbol: LOC100414470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744278.3 | XP_002744324.1 | 1 | 4122 | 93 | |
Set Membership: |
|