Assay Details
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744685.3 | XP_002744731.1 | 4 - 5 | 346 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744685.3 | XP_002744731.1 | 4 - 5 | 346 | 97 |
Set Membership: |
![]() ![]() ![]() |