Assay Details
| Gene Symbol: PNLIPRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756618.2 | XP_002756664.1 | 6 - 7 | 828 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PNLIPRP3| Gene Symbol: PNLIPRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756618.2 | XP_002756664.1 | 6 - 7 | 828 | 87 | |
Set Membership: |
|