Assay Details
| Gene Symbol: GLYAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755309.4 | XP_002755355.1 | 3 - 4 | 385 | 100 | |
| XM_009008168.1 | XP_009006416.1 | 3 - 4 | 363 | 100 | ||
| XM_009008169.1 | XP_009006417.1 | 3 - 4 | 363 | 100 | ||
More Information
Set Membership: |
|

GLYAT
Genomic Map