Assay Details
| Gene Symbol: SLC39A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759165.3 | XP_002759211.1 | 3 - 4 | 743 | 56 | |
| XM_008982997.2 | XP_008981245.1 | 2 - 3 | 457 | 56 | ||
| XM_017965504.1 | XP_017820993.1 | 3 - 4 | 778 | 56 | ||
More Information
Set Membership: |
|

SLC39A4
Genomic Map