Assay Details
| Gene Symbol: LGALS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760883.4 | XP_002760929.2 | 6 - 7 | 1126 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LGALS8| Gene Symbol: LGALS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760883.4 | XP_002760929.2 | 6 - 7 | 1126 | 121 | |
Set Membership: |
|