Assay Details
| Gene Symbol: DSPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732473.3 | XP_003732521.1 | 2 - 3 | 134 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSPP| Gene Symbol: DSPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732473.3 | XP_003732521.1 | 2 - 3 | 134 | 108 | |
Set Membership: |
|