Assay Details
| Gene Symbol: MSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979240.2 | XP_008977488.1 | 5 - 6 | 584 | 71 | |
| XM_008979241.2 | XP_008977489.1 | 5 - 6 | 581 | 71 | ||
| XM_017963740.1 | XP_017819229.1 | 3 - 4 | 431 | 71 | ||
More Information
Set Membership: |
|

MSR1
Genomic Map