Assay Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988551.2 | XP_008986799.1 | 4 - 5 | 439 | 96 | |
| XM_017968014.1 | XP_017823503.1 | 4 - 5 | 853 | 96 | ||
More Information
Set Membership: |
|

ZNF480
Genomic Map