Assay Details
| Gene Symbol: LOC100408200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963336.1 | XP_017818825.1 | 8 - 9 | 1070 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100408200| Gene Symbol: LOC100408200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963336.1 | XP_017818825.1 | 8 - 9 | 1070 | 91 | |