Assay Details
Gene Symbol: C5H17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971685.1 | XP_017827174.1 | 1 - 2 | 65 | 90 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C5H17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971685.1 | XP_017827174.1 | 1 - 2 | 65 | 90 |