Assay Details
| Gene Symbol: CST9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747513.3 | XP_002747559.1 | 2 - 3 | 1580 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST9L| Gene Symbol: CST9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747513.3 | XP_002747559.1 | 2 - 3 | 1580 | 73 | |