Assay Details
| Gene Symbol: DES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749815.4 | XP_002749861.1 | 7 - 8 | 1468 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DES| Gene Symbol: DES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749815.4 | XP_002749861.1 | 7 - 8 | 1468 | 70 | |
Set Membership: |
|