Assay Details
| Gene Symbol: KDELC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754367.2 | XP_002754413.1 | 1 - 2 | 257 | 88 | |
| XM_017977520.1 | XP_017833009.1 | 1 - 2 | 257 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KDELC2| Gene Symbol: KDELC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754367.2 | XP_002754413.1 | 1 - 2 | 257 | 88 | |
| XM_017977520.1 | XP_017833009.1 | 1 - 2 | 257 | 88 | ||