Assay Details
| Gene Symbol: BTNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806692.3 | XP_002806738.2 | 5 - 6 | 1820 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BTNL2| Gene Symbol: BTNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806692.3 | XP_002806738.2 | 5 - 6 | 1820 | 67 | |
Set Membership: |
|