Assay Details
| Gene Symbol: C2H5orf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965127.1 | XP_017820616.1 | 1 - 2 | 87 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2H5orf60| Gene Symbol: C2H5orf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965127.1 | XP_017820616.1 | 1 - 2 | 87 | 92 | |