Assay Details
| Gene Symbol: LOC100387423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971317.1 | XP_017826806.1 | 2 - 3 | 627 | 79 | |
| XM_017971320.1 | XP_017826809.1 | 2 - 3 | 627 | 79 | ||
More Information
Set Membership: |
|

LOC100387423
Genomic Map