Assay Details
| Gene Symbol: FOXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754469.3 | XP_002754515.2 | 2 - 3 | 375 | 86 | |
| XM_017977596.1 | XP_017833085.1 | 2 - 3 | 379 | 86 | ||
| XM_017977597.1 | XP_017833086.1 | 2 - 3 | 373 | 86 | ||
| XM_017977598.1 | XP_017833087.1 | 3 - 4 | 314 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017977598.1 transcripts.
Set Membership: |
|

FOXR1
Genomic Map