Assay Details
Gene Symbol: EFCAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758665.3 | XP_002758711.1 | 1 - 2 | 223 | 82 | |
XM_008982220.2 | XP_008980468.1 | 1 - 2 | 223 | 82 | ||
XM_008982223.2 | XP_008980471.1 | 1 - 2 | 223 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
