Assay Details
| Gene Symbol: WIPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998678.2 | XP_008996926.1 | 5 - 6 | 4388 | 68 | |
| XM_017973398.1 | XP_017828887.1 | 4 - 5 | 4133 | 68 | ||
| XM_017973399.1 | XP_017828888.1 | 4 - 5 | 562 | 68 | ||
More Information
Set Membership: |
|

WIPF1
Genomic Map