Assay Details
| Gene Symbol: LOC100895649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009869.2 | XP_009008117.2 | 1 | 825 | 95 | |
| XM_009009870.2 | XP_009008118.2 | 1 | 832 | 95 | ||
More Information
Set Membership: |
|

LOC100895649
Genomic Map