Assay Details
| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967344.1 | XP_017822833.1 | 4 - 5 | 505 | 77 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF546| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967344.1 | XP_017822833.1 | 4 - 5 | 505 | 77 | |