Assay Details
| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967344.1 | XP_017822833.1 | 5 - 6 | 625 | 99 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF546| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967344.1 | XP_017822833.1 | 5 - 6 | 625 | 99 | |