Assay Details
| Gene Symbol: ICAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967930.1 | XP_017823419.1 | 1 - 2 | 662 | 88 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ICAM3| Gene Symbol: ICAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967930.1 | XP_017823419.1 | 1 - 2 | 662 | 88 | |