Assay Details
| Gene Symbol: MPIG6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746349.2 | XP_002746395.1 | 1 - 2 | 98 | 110 | |
| XM_002746350.3 | XP_002746396.1 | 1 - 2 | 69 | 110 | ||
| XM_008994166.2 | XP_008992414.1 | 1 - 2 | 69 | 110 | ||
| XM_008994167.2 | XP_008992415.1 | 1 - 2 | 69 | 110 | ||
| XM_008994168.1 | XP_008992416.1 | 1 - 2 | 98 | 110 | ||
More Information
Set Membership: |
|

MPIG6B
Genomic Map