Assay Details
Gene Symbol: C11H11orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755189.3 | XP_002755235.1 | 2 - 3 | 303 | 96 | |
XM_009007945.1 | XP_009006193.1 | 2 - 3 | 288 | 96 | ||
XM_009007946.2 | XP_009006194.1 | 2 - 3 | 553 | 96 | ||
XM_009007947.2 | XP_009006195.1 | 3 - 4 | 294 | 96 |
