Assay Details
| Gene Symbol: CD247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985093.2 | XP_008983341.1 | 1 - 2 | 229 | 64 | |
| XM_008985094.2 | XP_008983342.1 | 1 - 2 | 234 | 64 | ||
More Information
Set Membership: |
|

CD247
Genomic Map