Assay Details
| Gene Symbol: LOC100411160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987451.2 | XP_008985699.1 | 1 - 2 | 1704 | 80 | |
| XM_017967878.1 | XP_017823367.1 | 2 - 3 | 1619 | 80 | ||
More Information
Set Membership: |
|

LOC100411160
Genomic Map