Assay Details
| Gene Symbol: LOC100390425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963436.1 | XP_017818925.1 | 8 - 9 | 828 | 75 | |
| XM_017963437.1 | XP_017818926.1 | 13 - 14 | 1620 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100390425| Gene Symbol: LOC100390425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963436.1 | XP_017818925.1 | 8 - 9 | 828 | 75 | |
| XM_017963437.1 | XP_017818926.1 | 13 - 14 | 1620 | 75 | ||