Assay Details
| Gene Symbol: LOC100388748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963981.1 | XP_017819470.1 | 5 - 6 | 1075 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100388748| Gene Symbol: LOC100388748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963981.1 | XP_017819470.1 | 5 - 6 | 1075 | 75 | |
Set Membership: |
|