Assay Details
| Gene Symbol: LOC103793080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996605.2 | XP_008994853.1 | 3 - 4 | 1491 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103793080| Gene Symbol: LOC103793080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996605.2 | XP_008994853.1 | 3 - 4 | 1491 | 66 | |
Set Membership: |
|