Assay Details
| Gene Symbol: LOC100394535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756370.2 | XP_002756416.1 | 2 - 3 | 139 | 90 | |
| XM_002756371.1 | XP_002756417.1 | 2 - 3 | 139 | 90 | ||
| XM_009009931.1 | XP_009008179.1 | 2 - 3 | 164 | 90 | ||
More Information
Set Membership: |
|

LOC100394535
Genomic Map