Assay Details
Gene Symbol: LOC100394535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756370.2 | XP_002756416.1 | 4 - 5 | 451 | 77 | |
XM_002756371.1 | XP_002756417.1 | 4 - 5 | 352 | 77 | ||
XM_009009931.1 | XP_009008179.1 | 4 - 5 | 476 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
