Assay Details
| Gene Symbol: AQP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753181.4 | XP_002753227.1 | 1 - 2 | 468 | 62 | |
| XM_009004961.2 | XP_009003209.1 | 1 - 2 | 468 | 62 | ||
| XM_017976610.1 | XP_017832099.1 | 1 - 2 | 468 | 62 | ||
More Information
Set Membership: |
|

AQP9
Genomic Map