Assay Details
| Gene Symbol: MST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758352.3 | XP_002758398.1 | 1 - 2 | 169 | 67 | |
| XM_008981640.2 | XP_008979888.1 | 1 - 2 | 111 | 67 | ||
More Information
Set Membership: |
|

MST1
Genomic Map