Assay Details
| Gene Symbol: PQLC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759462.2 | XP_002759508.1 | 2 - 3 | 323 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PQLC2L| Gene Symbol: PQLC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759462.2 | XP_002759508.1 | 2 - 3 | 323 | 67 | |
Set Membership: |
|