Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760352.4 | XP_002760398.1 | 1 - 2 | 39 | 66 | |
| XM_008984990.2 | XP_008983238.1 | 1 - 2 | 218 | 66 | ||
More Information
Set Membership: |
|

SERPINC1
Genomic Map