Assay Details
| Gene Symbol: C1H9orf129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993264.2 | XP_008991512.1 | 3 - 4 | 1126 | 81 | |
| XM_017972059.1 | XP_017827548.1 | 3 - 4 | 1123 | 81 | ||
More Information
Set Membership: |
|

C1H9orf129
Genomic Map