Assay Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963367.1 | XP_017818856.1 | 2 - 3 | 298 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

AVPI1| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963367.1 | XP_017818856.1 | 2 - 3 | 298 | 100 | |