Assay Details
| Gene Symbol: CAJA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746411.3 | XP_002746457.2 | 4 - 5 | 833 | 69 | |
| XM_008994228.1 | XP_008992476.1 | 3 - 4 | 922 | 69 | ||
| XM_008994229.1 | XP_008992477.1 | 4 - 5 | 790 | 69 | ||
More Information
Set Membership: |
|

CAJA-DOA
Genomic Map