Assay Details
| Gene Symbol: PTCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746624.3 | XP_002746670.2 | 1 - 2 | 596 | 69 | |
| XM_008994552.2 | XP_008992800.1 | 2 - 3 | 753 | 69 | ||
More Information
Set Membership: |
|

PTCHD4
Genomic Map